Chlorodimethyl-n-octadecylsilane, 95%
Chlorodimethyl-n-octadecylsilane, 95%
Chlorodimethyl-n-octadecylsilane, 95%
Thermo Scientific Chemicals

Chlorodimethyl-n-octadecylsilane, 95%

Preparation of hydrophobic surfaces, chain terminator | CAS: 18643-08-8 | C20H43ClSi | 347.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number 042414.14
also known as 042414-14
Price (USD)
81.65
Online Exclusive
90.80
Save 9.15 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
81.65
Online Exclusive
90.80
Save 9.15 (10%)
Each
Specifications
Beilstein2246012
Boiling Point184°C to 186°C (0.2mmHg)
CAS18643-08-8
Chemical Name or MaterialChlorodimethyl-n-octadecylsilane
ColorWhite
View more
Chlorodimethyl-n-octadecylsilane is used for preparation of hydrophobic surfaces and chain terminator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only