Boron, Oil based standard solution, Specpure™ B 5000μg/g
Thermo Scientific Chemicals

Boron, Oil based standard solution, Specpure™ B 5000μg/g

 
Have Questions?
產品號碼Quantity
042435.18
亦稱為 042435-18
50 g
產品號碼 042435.18
亦稱為 042435-18
價格 (TWD)
-
Quantity:
50 g
Request bulk or custom format
規格
Chemical Name or MaterialBoron, Oil based standard solution
MDL NumberMFCD00134034
Name NoteB 5000μg/g
Physical FormLiquid
Quantity50 g
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only