n-Hexacosane, 99%
n-Hexacosane, 99%
n-Hexacosane, 99%
Thermo Scientific Chemicals

n-Hexacosane, 99%

CAS: 630-01-3 | C26H54 | 366.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 042461.14
also known as 042461-14
Price (EUR)
96,70
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
96,70
Each
Add to cart
Chemical Identifiers
CAS630-01-3
IUPAC Namehexacosane
Molecular FormulaC26H54
InChI KeyHMSWAIKSFDFLKN-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormFlake
Appearance (Color)White waxy
Assay (GC)>98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only