Thermo Scientific Chemicals

n-Tetracosane, 99%, Thermo Scientific Chemicals

Catalog number: 042462.06
5 g, Each
Thermo Scientific Chemicals

n-Tetracosane, 99%, Thermo Scientific Chemicals

Catalog number: 042462.06
5 g, Each
Quantity
Catalog number: 042462.06
also known as 042462-06
Price (USD)
Price: 27.50
Online price: 23.65
Your price:
Quantity
-

Chemical Identifiers

CAS
646-31-1
Appearance (Color)
White
Assay (GC)
≥ 98.5%
Form
Waxy flake or powder

Description

n-Tetracosane is a suitable solvent used in the synthesis of ZnS nanoparticle. It is also employed as the wax component to study the reduction in pour point of hydrocarbon solvents containing wax crystals on addition of polymer additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text