n-Tetracosane, 99%
n-Tetracosane, 99%
n-Tetracosane, 99%
Thermo Scientific Chemicals

n-Tetracosane, 99%

CAS: 646-31-1 | C24H50 | 338.664 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 042462.14
also known as 042462-14
Price (EUR)
48,70
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
48,70
Each
Chemical Identifiers
CAS646-31-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥ 98.5%
FormWaxy flake or powder
n-Tetracosane is a suitable solvent used in the synthesis of ZnS nanoparticle. It is also employed as the wax component to study the reduction in pour point of hydrocarbon solvents containing wax crystals on addition of polymer additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only