Hafnium(IV) 2,4-pentanedionate, 97%
Hafnium(IV) 2,4-pentanedionate, 97%
Hafnium(IV) 2,4-pentanedionate, 97%
Hafnium(IV) 2,4-pentanedionate, 97%
Thermo Scientific Chemicals

Hafnium(IV) 2,4-pentanedionate, 97%

CAS: 17475-67-1 | C20H28HfO8 | 574.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042463.06
also known as 042463-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS17475-67-1
IUPAC Namehafnium(4+) tetrakis(2,4-dioxopentan-3-ide)
Molecular FormulaC20H28HfO8
InChI KeyGXNMJQUZCICHBX-UHFFFAOYSA-N
SMILES[Hf+4].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystals or crystalline powder
Assay from Suppliers CofAHf: ≥99.5% (metals basis)
Elemental Analysis≥96%
Elemental AnalysisC: 40.7- 42.8%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only