Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 770
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 770
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 770
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 770
Thermo Scientific Chemicals

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 770

CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Catalog number 042497.AK
also known as 042497-AK
Price (EUR)
101,00
Each
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
101,00
Each
Add to cart
Chemical Identifiers
CAS9016-00-6
Molecular Formula(C2H6OSi)n
SMILESC[Si](C)(-*)O-*
MDL NumberMFCD00084411
SpecificationsSpecification SheetSpecification Sheet
FormViscous liquid
Appearance (Color)Colorless
Infrared spectrumConforms to structure
Viscosity4-6 cSt
Polydimethylsiloxane, trimethylsiloxy is used in coating Auxiliary Agents. It is also used as Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only