Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 28,000
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 28,000
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 28,000
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 28,000
Thermo Scientific Chemicals

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 28,000

CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Catalog number 042503.AK
also known as 042503-AK
Price (EUR)
57,00
Each
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
57,00
Each
Add to cart
Chemical Identifiers
CAS9016-00-6
Molecular Formula(C2H6OSi)n
SMILESC[Si](C)(-*)O-*
MDL NumberMFCD00084411
SpecificationsSpecification SheetSpecification Sheet
Viscosity900-1100 cSt (25°C)
Appearance (Color)Colorless
Infrared spectrumConforms
FormViscous liquid

Polydimethylsiloxane, trimethylsiloxy is used in water-repellent, protective creams & lotions. It is also used as surfactant, is used in coating auxiliary agents. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only