Glassy carbon spherical powder, 200-400 micron, type 2
Thermo Scientific Chemicals

Glassy carbon spherical powder, 200-400 micron, type 2

Used as such in purifying metals, in electrodes, electrical devices & steel. Glassy Carbon Spherical Powder has numerous applications due its fundamental softness, including in lubricant formulations and as an additive to structural materials and metallic alloys.
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 042550.09
also known as 042550-09
Price (USD)
64.65
Special offer
Online exclusive
Ends: 28-Dec-2026
75.40
Save 10.75
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
64.65
Special offer
Online exclusive
Ends: 28-Dec-2026
75.40
Save 10.75
Each
Specifications
Chemical Name or MaterialGlassy carbon spherical powder
DOT InformationHazard Class: 4.1; Packaging Group: III
Health Hazard 1H228
Health Hazard 3P210-P240-P241-P280-P370+P378q
MDL NumberMFCD00133992
View more

Glassy carbon spherical powder is used as such in purifying metals, in electrodes, electrical devices & steel. Glassy Carbon Spherical Powder has numerous applications due its fundamental softness, including in lubricant formulations and as an additive to structural materials and metallic alloys.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

What is the difference between type 1 glassy carbon and type 2 glassy carbon?

Type 1 is a "low temperature" glassy carbon produced by heat treatment at ~ 1,000-2,000°C and consists of discreet fragments of distorted graphene layers. In general, type 1 is less compressible, more resistant to oxidation, and less permeable to gasses. Type 2 is a"high temperature" glassy carbon produced by heat treatment at ~2,500-3,000°C and consists of broken or imperfect fullerene-like nanospheroids encased in a disordered, multilayered graphene matrix. Type 2 usually contains fewer impurities and has higher electrical and thermal conductivity.