Dichromate, Ion chromatography standard solution, Specpure&trade;, Cr2O7<sup>-2</sup> 1000&mu;g/mL
Thermo Scientific Chemicals

Dichromate, Ion chromatography standard solution, Specpure™, Cr2O7-2 1000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 042565.AP
also known as 042565-AP
Price (TWD)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialDichromate, Ion chromatography standard solution
Health Hazard 1H317-H332-H334-H340-H350-H360
Health Hazard 2GHS H Statement
H340-H350-H332
May cause genetic defects.
May cause cancer.
Harmful if inhaled.
Health Hazard 3P201-P202-P261-P271-P272-P280g-P281-P285-P302+P352-P304+P340-P308+P313-P333+P313-P342+P311-P363-P501c
MDL NumberMFCD01093393
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only