Iodide, Ion chromatography standard solution, Specpure™ I∣- 1000μg/mL
Thermo Scientific Chemicals

Iodide, Ion chromatography standard solution, Specpure™ I∣- 1000μg/mL

Have Questions?
變更視圖buttonViewtableView
Quantity:
100 mL
500 mL
產品號碼 042567.AP
價格 (TWD)
-
Quantity:
500 mL
Request bulk or custom format
規格
Chemical Name or MaterialIodide, Ion chromatography standard solution
MDL NumberMFCD00011355
Name NoteI- 1000μg/mL
Physical FormLiquid
Quantity500 mL
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only