Thermo Scientific Chemicals

Aluminum oxide, Alpha-phase, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 042572.36
500 g, Each
Thermo Scientific Chemicals

Aluminum oxide, Alpha-phase, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 042572.36
500 g, Each
Quantity
Catalog number: 042572.36
also known as 042572-36
Price (USD)

Chemical Identifiers

CAS
1344-28-1
IUPAC Name
dialuminium(3+) trioxidandiide
Molecular Formula
Al2O3
InChI Key
PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Al+3].[Al+3]
Form
Powder
Appearance (Color)
White
Comment
Crystal structure: Alpha phase
Particle Size
APS, D₅₀: typically 0.5-0.7µm
Total Metal Impurities
≤0.1% (≤1,000ppm)

Description

Aluminum oxide is widely used in a variety of plastics, rubber, ceramics, refractory products for reinforcement toughening. It is also useful to produce artificial ruby, sapphire, yttrium aluminum garnet. It is useful in the field of fiber fabric products and high pressure sodium lamp as far-infrared emission and thermal insulation materials. It is useful for the C single crystal detectors. It plays an important role for the hydrogenation of aromatic hydrocarbons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text