Palladium hydroxide, Pd 20% on carbon, nominally 50% water, Pearlman's Catalyst
Palladium hydroxide, Pd 20% on carbon, nominally 50% water, Pearlman's Catalyst
Palladium hydroxide, Pd 20% on carbon, nominally 50% water, Pearlman's Catalyst
Thermo Scientific Chemicals

Palladium hydroxide, Pd 20% on carbon, nominally 50% water, Pearlman's Catalyst

CAS: 12135-22-7 | H2O2Pd | 140.43 g/mol
Have Questions?
Catalog NumberQuantity
042578.04
also known as 042578-04
2 g
Catalog number 042578.04
also known as 042578-04
Price (EUR)
168,00
Each
Quantity:
2 g
Request bulk or custom format
Price (EUR)
168,00
Each
Chemical Identifiers
CAS12135-22-7
IUPAC Namepalladium(2+) dihydroxide
Molecular FormulaH2O2Pd
InChI KeyNXJCBFBQEVOTOW-UHFFFAOYSA-L
SMILES[OH-].[OH-].[Pd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Pd 18.00-22.00%
CommentMetal area 23.0% min
CommentNitrobenzene activity 150 min
Assay (unspecified)Moisture 40.00-60.00%

Hydrogenolysis of benzyl-nitrogen bondsPalladium hydroxide, Pd 20% on carbon, nominally 50% water is used as a catalyst for hydrogenolysis of benzyl groups and epoxides. It is used to reduce nitro compounds to amines. It acts as a dehydrogenation reagent as well as a useful reagent for debenzylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Used as catalyst for hydrogenolysis of benzyl groups and epoxides.
It is used to reduce nitro compounds to amines.
Acts as dehydrogenation reagent as well as useful reagent for debenzylation
RUO – Research Use Only