Thermo Scientific™
Ground & Waste Water Pollution Standard Solution 5, Specpure™, Thermo Scientific™
100mL, Each
Thermo Scientific™
Ground & Waste Water Pollution Standard Solution 5, Specpure™, Thermo Scientific™
100mL, Each
Quantity
Catalog number: 042580.AE
also known as 042580-AE
Price (USD)
118.00 / Each
Your Price:
Quantity
-
Concentration
Mg 100?g/mL ?2%
Concentration
Na 500?g/mL ?2%
Comment
For US EPA Clean Water Act.
Concentration
Ca 500?g/mL ?2%
Concentration
K 100?g/mL ?2%
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.