Thermo Scientific Chemicals

Azelaic acid, 96%, Thermo Scientific Chemicals

Catalog number: 042587.36
500 g, Each
Thermo Scientific Chemicals

Azelaic acid, 96%, Thermo Scientific Chemicals

Catalog number: 042587.36
500 g, Each
Quantity
Catalog number: 042587.36
also known as 042587-36
Price (USD)
Price: 214.00
Online price: 184.65
Your price:
Quantity
-

Description

Azelaic acid acts as an anti-inflammatory compound. It is also exhibits anti acne activity. It up regulates expression of ant oxidative enzymes. Azelaic acid also inhibits proliferation of melanocytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text