Di-n-octyldichlorosilane, 97%
Di-n-octyldichlorosilane, 97%
Di-n-octyldichlorosilane, 97%
Thermo Scientific Chemicals

Di-n-octyldichlorosilane, 97%

CAS: 18416-07-4 | C16H34Cl2Si | 325.433 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 042599.09
also known as 042599-09
Price (USD)
93.60
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
93.60
Each
Chemical Identifiers
CAS18416-07-4
IUPAC Namedichlorodioctylsilane
Molecular FormulaC16H34Cl2Si
InChI KeyCVAGYCLEIYGJQT-UHFFFAOYSA-N
SMILESCCCCCCCC[Si](Cl)(Cl)CCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)> 96.0%
Di-n-octyldichlorosilane is used in the synthesis of organic photovoltaics. It is also used in medicine field, electronic industry and polymer material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only