Fullerene powder, 99+% C{70}
Fullerene powder, 99+% C{70}
Fullerene powder, 99+% C{70}
Thermo Scientific Chemicals

Fullerene powder, 99+% C{70}

CAS: 115383-22-7 | C70 | 840.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 042600.MD
also known as 042600-MD
Price (TWD)
-
Quantity:
250 mg
Request bulk or custom format
Specifications
Assay Percent NotesC70
CAS115383-22-7
ChEBICHEBI:33195
Chemical Name or MaterialFullerene powder
ColorGray to Black
View more
Fullerene powder is used for biomedical applications including the design of high-performance magnetic resonance imaging (MRI) contrast agents, X-ray imaging contrast agents, photodynamic therapy and drug and gene delivery. It is used in superconductors, photoactive polymers, film material modifiers, abrasive agent for cutting and grinding applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only