Thermo Scientific Chemicals

Cobalt foil, 0.1mm (0.004in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

100 x 100 mm, Each
Thermo Scientific Chemicals

Cobalt foil, 0.1mm (0.004in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

100 x 100 mm, Each
Quantity
Catalog number: 042658.GH
also known as 042658-GH
Price (USD)
Price: 480.00
Your Price:
Quantity
-

Chemical Identifiers

CAS
7440-48-4
IUPAC Name
cobalt
Molecular Formula
Co
InChI Key
GUTLYIVDDKVIGB-UHFFFAOYSA-N
SMILES
[Co]
Appearance (Color)
Blue silver
Form
Foil, 0.1mm (0.004in) thick.
Assay from Supplier's CofA
≥99.95% (metals basis)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only