Dihydroxybis(ammonium lactato)titanium(IV), 50% w/w aq. soln.
Dihydroxybis(ammonium lactato)titanium(IV), 50% w/w aq. soln.
Dihydroxybis(ammonium lactato)titanium(IV), 50% w/w aq. soln.
Thermo Scientific Chemicals

Dihydroxybis(ammonium lactato)titanium(IV), 50% w/w aq. soln.

Water soluble titanium complex | CAS: 65104-06-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number 042676.30
also known as 042676-30
Price (USD)
46.65
Special offer
Online exclusive
Ends: 30-Jun-2026
54.40
Save 7.75 (14%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
46.65
Special offer
Online exclusive
Ends: 30-Jun-2026
54.40
Save 7.75 (14%)
Each
Chemical Identifiers
CAS65104-06-5
SpecificationsSpecification SheetSpecification Sheet
Specific Gravity1.180-1.250 @ 25?C
Appearance (Color)Clear to turbid colorless to yellow
Assay from Supplier's CofATi: 13.20-14.00% (as TiO2)
Chloride content≤120ppm
Color (Gardner Scale)≤6
View more
Dihydroxybis(ammonium lactato)titanium(IV) is used in the production of water soluble titanium complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only