Zinc wire, 1.0mm (0.04in) dia, 99.95% (metals basis)
Zinc wire, 1.0mm (0.04in) dia, 99.95% (metals basis)
Zinc wire, 1.0mm (0.04in) dia, 99.95% (metals basis)
Zinc wire, 1.0mm (0.04in) dia, 99.95% (metals basis)
Thermo Scientific Chemicals

Zinc wire, 1.0mm (0.04in) dia, 99.95% (metals basis)

CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
25 m
Catalog number 042705.G9
also known as 042705-G9
Price (EUR)
342,00
Each
Quantity:
25 m
Request bulk or custom format
Price (EUR)
342,00
Each
Specifications
Assay Percent Notes(metals basis)
CAS7440-66-6
ChEBICHEBI:30185
Chemical Name or MaterialZinc wire
EINECS Number231-175-3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only