Tetramethylammonium formate, 30% w/w aq. soln.
Tetramethylammonium formate, 30% w/w aq. soln.
Tetramethylammonium formate, 30% w/w aq. soln.
Thermo Scientific Chemicals

Tetramethylammonium formate, 30% w/w aq. soln.

CAS: 59138-84-0 | C5H13NO2 | 119.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 042710.AE
also known as 042710-AE
Price (USD)
88.30
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
88.30
Each
Chemical Identifiers
CAS59138-84-0
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Suppliers CofA29.0-31.0%
Tetramethylammonium formate may provide the lowest temperature sensitivity, additionally reducing corrosion of stainless-steel flow system by chloride-induced depassivation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only