Polystyrene latex microsphere, 0.10 micron, 2.5 wt% dispersion in water
Polystyrene latex microsphere, 0.10 micron, 2.5 wt% dispersion in water
Polystyrene latex microsphere, 0.10 micron, 2.5 wt% dispersion in water
Thermo Scientific Chemicals

Polystyrene latex microsphere, 0.10 micron, 2.5 wt% dispersion in water

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol
Have Questions?
Catalog NumberQuantity
042712.4Y
also known as 042712-4Y
15 mL
Catalog number 042712.4Y
also known as 042712-4Y
Price (EUR)
447,65
Online Exclusive
497,00
Save 49,35 (10%)
Each
Quantity:
15 mL
Request bulk or custom format
Price (EUR)
447,65
Online Exclusive
497,00
Save 49,35 (10%)
Each
Chemical Identifiers
CAS9003-53-6
Molecular Formula(C8H8)n
SMILES*-CC(-*)C1=CC=CC=C1
MDL NumberMFCD00084450
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)0 to 15% ( CV/Coefficient of variance:)
Particle Size0.079 To 0.121 Micron
Appearance (Color)White
FormLiquid
Assay (unspecified)solids content: 2.5 to 2.8 wt%
Polystyrene latex microsphere is covalently attached to a single-stranded DNA in order to increase the charge stabilization of the microspheres. Further, it is used as an efficient support for immunological reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only