Thermo Scientific Chemicals

Silica gel, wide pore, 150 angstroms

Catalog number: 042726.A1
1 kg, Each
Thermo Scientific Chemicals

Silica gel, wide pore, 150 angstroms

Catalog number: 042726.A1
1 kg, Each
Quantity
Catalog number: 042726.A1
also known as 042726-A1
Price (USD)
Quantity
-

Chemical Identifiers

CAS
63231-67-4
IUPAC Name
dihydroxysilanone
Molecular Formula
H2O3Si
InChI Key
IJKVHSBPTUYDLN-UHFFFAOYSA-N
SMILES
O[Si](O)=O
Appearance (Color)
White
Form
Powder
Loss on Drying
≤2.0%
Particle Size
Pore diameter (av.): 130-170 angstroms
Particle Size
Report particle size distribution (microns, typical 40-75)

Description

Silica gel, wide pore is mainly used for the dehydrated purification of the industrial gases, the clearance of the organic acids and high polymers in the insulative oil, the purification and separation of the medicines. It is also used in column chromatography without applied pressure (gravity pulled).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text