Thermo Scientific Chemicals

Silica gel 60, 0.019-0.037mm (400-600 mesh), Thermo Scientific Chemicals

Catalog number: 042729.A1
1 kg, Each
Thermo Scientific Chemicals

Silica gel 60, 0.019-0.037mm (400-600 mesh), Thermo Scientific Chemicals

Catalog number: 042729.A1
1 kg, Each
Quantity
Catalog number: 042729.A1
Price (USD)
Quantity
-

Chemical Identifiers

CAS
7631-86-9
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Appearance (Color)
White to pale gray
Particle Size
18-37 Microns
Particle Size
Mean Pore Diameter: 60 Angstroms
Surface Area
500-600 m²/g
Form
Powder

Description

Column chromatography with elevated pressureSilica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text