Polystyrene latex microsphere, 25.0 micron, 2.5 wt% dispersion in water
Polystyrene latex microsphere, 25.0 micron, 2.5 wt% dispersion in water
Polystyrene latex microsphere, 25.0 micron, 2.5 wt% dispersion in water
Polystyrene latex microsphere, 25.0 micron, 2.5 wt% dispersion in water
Thermo Scientific Chemicals

Polystyrene latex microsphere, 25.0 micron, 2.5 wt% dispersion in water

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol
Have Questions?
Catalog NumberQuantity
042730.AA
also known as 042730-AA
5 mL
Catalog number 042730.AA
also known as 042730-AA
Price (USD)
568.65
Special offer
Online exclusive
Ends: 30-Jun-2026
669.00
Save 100.35 (15%)
Each
Add to cart
Quantity:
5 mL
Request bulk or custom format
Price (USD)
568.65
Special offer
Online exclusive
Ends: 30-Jun-2026
669.00
Save 100.35 (15%)
Each
Add to cart
Specifications
CAS9003-53-6
ChEBICHEBI:27452
Chemical Name or MaterialPolystyrene latex microsphere
Concentration or Composition (by Analyte or Components)2.5 wt% dispersion in water
IUPAC Namestyrene
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only