Silicon(IV) oxide, Amorphous fumed, S.A. 300 to 350m2/g
Silicon(IV) oxide, Amorphous fumed, S.A. 300 to 350m2/g
Silicon(IV) oxide, Amorphous fumed, S.A. 300 to 350m2/g
Thermo Scientific Chemicals

Silicon(IV) oxide, Amorphous fumed, S.A. 300 to 350m2/g

For Adhesives, Coatings, Cosmetics | CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Catalog NumberQuantity
042741.30
also known as 042741-30
250 g
Catalog number 042741.30
also known as 042741-30
Price (EUR)
97,70
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
97,70
Each
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Surface Area300-350m²/g
Adhesives, coatings, cosmetics, defomers, inks, lubricants, sealants, thermal insulationSilicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of food and pharmaceutical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

S.A. 300 to 350m2/g
RUO – Research Use Only