Dihydrogen hexachloroplatinate(IV) hexahydrate, low nitrate, Premion™, 99.95% (metals basis), Pt 37.5% min
Dihydrogen hexachloroplatinate(IV) hexahydrate, low nitrate, Premion™, 99.95% (metals basis), Pt 37.5% min
Dihydrogen hexachloroplatinate(IV) hexahydrate, low nitrate, Premion™, 99.95% (metals basis), Pt 37.5% min
Thermo Scientific Chemicals

Dihydrogen hexachloroplatinate(IV) hexahydrate, low nitrate, Premion™, 99.95% (metals basis), Pt 37.5% min

CAS: 18497-13-7 | Cl6H14O6Pt | 517.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 042815.14
also known as 042815-14
Price (EUR)
1.898,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
1.898,00
Each
Chemical Identifiers
CAS18497-13-7
IUPAC Nameplatinum(4+) dihydrogen hexahydrate hexachloride
Molecular FormulaCl6H14O6Pt
InChI KeyPIJUVEPNGATXOD-UHFFFAOYSA-J
SMILES[H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]
View more
SpecificationsSpecification SheetSpecification Sheet
FormLump
Appearance (Color)Orange
Assay (special)Pt 37.5-39.9%
Total Metal Impurities≤0.05%
Dihydrogen hexachloroplatinate(IV) hexahydrate is used for the determination of potassium. It is also used for the purification of platinum by treating it with ammonium chloride flowed by heating of ammonium hexachloroplatinate with hydrogen yields elemental platinum. It also finds application in catalysis, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink and microscopy. It is a precursor for the preparation of other platinum catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only