Ytterbium(III) chloride, 35% min w/w aq. soln., 99.9% (REO)
Ytterbium(III) chloride, 35% min w/w aq. soln., 99.9% (REO)
Ytterbium(III) chloride, 35% min w/w aq. soln., 99.9% (REO)
Thermo Scientific Chemicals

Ytterbium(III) chloride, 35% min w/w aq. soln., 99.9% (REO)

CAS: 10361-91-8 | YbCl3
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Catalog number 042816.AD
also known as 042816-AD
Price (USD)
55.90
Each
Quantity:
50 mL
Request bulk or custom format
Price (USD)
55.90
Each
Chemical Identifiers
CAS10361-91-8
IUPAC Nameytterbium(3+) trichloride
Molecular FormulaCl3Yb
InChI KeyCKLHRQNQYIJFFX-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Yb+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Applied with NiCl2 to form a very effective catalyst for the reductive dehalogenation of aryl halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only