Manganese(II) chloride, ultra dry, 99.99% (metals basis)
Manganese(II) chloride, ultra dry, 99.99% (metals basis)
Manganese(II) chloride, ultra dry, 99.99% (metals basis)
Thermo Scientific Chemicals

Manganese(II) chloride, ultra dry, 99.99% (metals basis)

Plays an important role in the production of dry cell batteries. | CAS: 7773-01-5 | Cl2Mn | 125.84 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
2 g
10 g
50 g
產品號碼 042844.09
亦稱為 042844-09
價格 (TWD)
-
Quantity:
10 g
Request bulk or custom format
化學識別
CAS7773-01-5
IUPAC Namemanganese(2+) dichloride
Molecular FormulaCl2Mn
InChI KeyGLFNIEUTAYBVOC-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Mn++]
檢視更多
規格Specification Sheet規格表
Residual water< 100 ppm
Total Metal Impurities0.01% max.
FormPowder
Appearance (Color)Pink
Manganese(II) chloride, plays an important role in the production of dry cell batteries. It is also used as a starting material to the methylcyclopentadienyl manganese tricarbonyl, which has antiknock property. Further, it is used in the formation of an active Manganese(0) species useful for radical cyclization. It is also used in 31P-NMR to determine the size and lamellarity of phospholipidvesicles reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only