Thermo Scientific Chemicals

Bismuth(III) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 042863.30
250 g, Each
Thermo Scientific Chemicals

Bismuth(III) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 042863.30
250 g, Each
Quantity
Catalog number: 042863.30
also known as 042863-30
Price (USD)
Quantity
-

Chemical Identifiers

CAS
1304-76-3
IUPAC Name
dibismuth(3+) trioxidandiide
Molecular Formula
Bi2O3
InChI Key
TYIXMATWDRGMPF-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Bi+3].[Bi+3]
Total Metal Impurities
0.001% max.

Description

Bismuth(III) oxide is used in solid-oxide fuel cells. It is also used in optical glasses, to replace lead oxide in whitewares, luxes, varistor formulations and ceramic capacitor formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text