Thermo Scientific Chemicals
Platinum 6 wt% Rhodium wire, 0.25mm (0.01in) dia, ISA Type B Standard Grade Thermocouple, Thermo Scientific Chemicals
Catalog number: 042913.BY
100 cm, Each
Thermo Scientific Chemicals
Platinum 6 wt% Rhodium wire, 0.25mm (0.01in) dia, ISA Type B Standard Grade Thermocouple, Thermo Scientific Chemicals
Catalog number: 042913.BY
100 cm, Each
Quantity
Catalog number: 042913.BY
also known as 042913-BY