Lithium isopropoxide, 99.9% (metals basis)
Lithium isopropoxide, 99.9% (metals basis)
Lithium isopropoxide, 99.9% (metals basis)
Thermo Scientific Chemicals

Lithium isopropoxide, 99.9% (metals basis)

CAS: 2388-10-5 | C3H7LiO | 66.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 042930.09
also known as 042930-09
Price (EUR)
291,00
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
291,00
Each
Specifications
Assay Percent Notes(metals basis)
Beilstein3618614
CAS2388-10-5
Chemical Name or MaterialLithium isopropoxide
ColorWhite to Yellow
View more
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only