Germanium rod, polycrystalline, 6.35mm (0.25in) dia, 99.9999% (metals basis)
Germanium rod, polycrystalline, 6.35mm (0.25in) dia, 99.9999% (metals basis)
Germanium rod, polycrystalline, 6.35mm (0.25in) dia, 99.9999% (metals basis)
Germanium rod, polycrystalline, 6.35mm (0.25in) dia, 99.9999% (metals basis)
Thermo Scientific Chemicals

Germanium rod, polycrystalline, 6.35mm (0.25in) dia, 99.9999% (metals basis)

CAS: 7440-56-4 | Ge | 72.63 g/mol
Have Questions?
Catalog NumberQuantity
042951.BS
also known as 042951-BS
10 cm
Catalog number 042951.BS
also known as 042951-BS
Price (USD)/ Each
687.65
Online Exclusive
764.00
Save 76.35 (10%)
Add to cart
Quantity:
10 cm
Request bulk or custom format
Price (USD)/ Each
687.65
Online Exclusive
764.00
Save 76.35 (10%)
Add to cart
Specifications
Assay Percent Notes(metals basis)
CAS7440-56-4
ChEBICHEBI:30441
Chemical Name or MaterialGermanium rod
EINECS Number231-164-3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only