Dionex™ Carbohydrate Standards
Actual product may vary
Thermo Scientific™

Dionex™ Carbohydrate Standards

Enable speedy performance verification of your carbohydrate system with these ready-made Thermo Scientific™ Dionex™ Carbohydrate Standards.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
043064OligoStandard
043162MonoStandard
Catalog number 043064
Price (MXN)
-
Type:
OligoStandard
Use these two ready-made, qualitative Thermo Scientific™ Dionex™ Carbohydrate Standards to enable speedy performance verification of your Thermo Scientific™ BioLC™ system or CarboPac™ carbohydrate columns. Simply reconstitute the vial contents and inject, for fast and easy verification.
Product Detail

Dionex™ MonoStandard™ Mix of Six

The Dionex MonoStandard Mix of Six carbohydrate standards is prepared from reference-grade monosaccharides. Prior to use, dilute the contents of the vial to yield a solution containing 0.1mM (100pmol/μL) of each monosaccharide.

Each vial of Dionex MonoStandard Mix of Six contains:

  • 100nmol fucose
  • 100nmol galactosamine hydrochloride
  • 100nmol glucosamine hydrochloride
  • 100nmol galactose
  • 100nmol glucose
  • 100nmol mannose

Dionex Oligo Standard

The Dionex Oligo Standard is purified from bovine fetuin. Prior to use, reconstitute the contents of the vial in a known volume of water (e.g. 250mL) to suit your application.

Each vial of Dionex Oligo Standard contains the following oligosaccharides;

  • 25nmol sialylated fetuin N-linked alditols

Quality to Trust In

All Thermo Scientific Dionex IC standards are traceable to National Institute of Standards and Technology (NIST) Standard Reference Materials and are shipped with a Certificate of Analysis verifying the concentration.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Concentration25 nmol
DescriptionDionex OligoStandard, Sialylated Fetuin N-Linked Alditols
For Use With (Equipment)BioLC Systems; CarboPac Carbohydrate Columns
Quantity25 nmol
Standard ComponentsSialylated N-linked Fetuin Alditols
Standard Concentration25 nmol
TypeOligoStandard
Unit SizeEach 1