Thermo Scientific Chemicals

Drierite™, indicating (CaSO{4}), Thermo Scientific Chemicals

Catalog number: 043064.P2
1 lb., Each
Thermo Scientific Chemicals

Drierite™, indicating (CaSO{4}), Thermo Scientific Chemicals

Catalog number: 043064.P2
1 lb., Each
Quantity
Catalog number: 043064.P2
also known as 043064-P2
Price (USD)
Price: 46.60
Online price: 39.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7778-18-9
IUPAC Name
calcium sulfate
Molecular Formula
CaO4S
InChI Key
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
SMILES
[Ca++].[O-]S([O-])(=O)=O
Appearance (Color)
Blue when active, pink when exhausted
Form
Granular, irregular, -6 mesh
Assay from Supplier's CofA
Mositure: 0.0-0.3%

Description

Drierite™, indicating (CaSO{4}) is used as a desiccant. It is used as a drying agent utilized for the efficient and rapid drying of air, industrial gases, refrigerants, organic liquids and solids. Further, it is used to maintain a dry atmosphere in storage spaces, vaults, commercial packages, and other enclosures. In addition to this, it is involved in the protection of hygroscopic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text