Silicon(IV) oxide, 30% in Ethylene glycol, colloidal dispersion, Thermo Scientific Chemicals
Silicon(IV) oxide, 30% in Ethylene glycol, colloidal dispersion, Thermo Scientific Chemicals
Silicon(IV) oxide, 30% in Ethylene glycol, colloidal dispersion, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silicon(IV) oxide, 30% in Ethylene glycol, colloidal dispersion, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
043093.A1
also known as 043093-A1
1 kg
Catalog number 043093.A1
also known as 043093-A1
Price (EUR)
79,65
Online exclusive
87,90
Save 8,25 (9%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
79,65
Online exclusive
87,90
Save 8,25 (9%)
Each
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size45-95nm
Concentration (weight/weight)≥30%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only