Thermo Scientific Chemicals

5-Sulfosalicylic acid dihydrate, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 043144.14
25 g, Each
Thermo Scientific Chemicals

5-Sulfosalicylic acid dihydrate, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 043144.14
25 g, Each
Quantity
Catalog number: 043144.14
also known as 043144-14
Price (USD)
Price: 32.60
Online price: 25.65
Your price:
Quantity
-

Chemical Identifiers

CAS
5965-83-3
IUPAC Name
2-hydroxy-5-sulfobenzoic acid; bis(methane)
Molecular Formula
C9H14O6S
InChI Key
NFYHZVWMQHQKRU-UHFFFAOYSA-N
SMILES
C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
99.0 - 101.0% ( as a Dihydrate)
Form
solid
Chloride content
0.001%, max
Impurity content
Salicylic acid: 0.04%, max

Description

Reagent for precipitation of proteins5-Sulfosalicylic acid dihydrate is used as a metal scavenger and it forms proton-transfer dye complexes with diazo compounds such as 4-(phenyldiazenyl)aniline. It is employed as a preparative measure for removing proteins prior to chromatographic analysis. It plays a vital role in reducing and fixation of proteins in agarose and polyacrylamide gels. It acts as a surfactant and lubricating grease additive. It is also used in urine tests to determine urine protein content. Further, it acts as an intermediate in the synthesis of active pharmaceutical ingredients like doxycycline and methacycline. In addition to this, it is used as a spray reagent for the detection of amino acids on thin layer chromatography (TLC) plates and as a doping agent for polyaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text