Lithium niobium isopropoxide, 99+% (metals basis), 5% w/v in isopropanol, Thermo Scientific Chemicals
Lithium niobium isopropoxide, 99+% (metals basis), 5% w/v in isopropanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium niobium isopropoxide, 99+% (metals basis), 5% w/v in isopropanol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Catalog number 043175.AE
also known as 043175-AE
Price (USD)/ Each
263.00
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
263.00
Add to cart
Lithium niobium isopropoxide, 99+% (metals basis), 5% w/v in isopropanol, Thermo Scientific Chemicals
Catalog number043175.AE
Price (USD)/ Each
263.00
-
Add to cart
Specifications
Chemical Name or MaterialLithium niobium isopropoxide
CAS21864-21-1
Health Hazard 1H225-H314-H335-H336
Health Hazard 2GHS H Statement
H225-H315-H319-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
View more
Used as solid-state electrolyte.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only