Ammonium dichromate, 99.995% (metals basis)
Ammonium dichromate, 99.995% (metals basis)
Ammonium dichromate, 99.995% (metals basis)
Thermo Scientific Chemicals

Ammonium dichromate, 99.995% (metals basis)

CAS: 7789-09-5 | Cr2H8N2O7 | 252.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 043180.14
also known as 043180-14
Price (EUR)
165,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
165,00
Each
Chemical Identifiers
CAS7789-09-5
IUPAC Namediammonium [(oxidodioxochromio)oxy]chromiumoylolate
Molecular FormulaCr2H8N2O7
InChI KeyJOSWYUNQBRPBDN-UHFFFAOYSA-P
SMILES[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red
FormPowder or crystals or crystalline powder
Total Metal Impurities≤0.005%
Ammonium dichromate is used in photo engraving, dye mordant, pigments, pickling, photography, process engraving, lithography, pyrotechnics, glazes for porcelain and china wares. It acts as a catalyst and a source of nitrogen in the laboratory and as the photoactive site. It is also used in the preparation of alizarin and chrome alum. Further, it is used in leather and oil industry for tanning and purification respectively. It is involved in the production of the phosphor raster television screens and other devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only