Acetyl chloride, 99+%
Acetyl chloride, 99+%
Acetyl chloride, 99+%
Thermo Scientific Chemicals

Acetyl chloride, 99+%

CAS: 75-36-5 | C2H3ClO | 78.50 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Catalog number 043262.K2
also known as 043262-K2
Price (EUR)
232,65
Online Exclusive
258,00
Save 25,35 (10%)
Each
Quantity:
1 L
Request bulk or custom format
Price (EUR)
232,65
Online Exclusive
258,00
Save 25,35 (10%)
Each
Chemical Identifiers
CAS75-36-5
IUPAC Nameacetyl chloride
Molecular FormulaC2H3ClO
InChI KeyWETWJCDKMRHUPV-UHFFFAOYSA-N
SMILESCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
FormLiquid
Acetyl chloride acts as a reagent for the preparation of esters and amides of acetic acid. It is also useful an important reactant in Friedel-Crafts reactions as well as the introduction of an acetyl group. It serves as a starting material in the production of pharmaceutical, new plating complexing agent, acylation agent and synthetic organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only