Control Sets | MGC Primary DAU Control Set |
Description | DRI Propoxyphene Drugs of Abuse Assay |
Detectable Analytes | Propoxyphene |
DoA Calibrators | DRI Multi-Drug Calibrators |
Sample Type | Urine |
Type | Reagent |
Quantity | 1 x 500 mL |
Unit Size | Each |
Control Sets | MGC Primary DAU Control Set |
Description | DRI Propoxyphene Drugs of Abuse Assay |
Detectable Analytes | Propoxyphene |
DoA Calibrators | DRI Multi-Drug Calibrators |
Sample Type | Urine |
Type | Reagent |
Quantity | 1 x 100 mL |
Unit Size | Each |
Detectable Analytes | Propoxyphene |
Sample Type | Urine |
Type | Reagent |
Quantity | 3 x 18 mL |
Shelf Life | Until Label Expiry Date |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
0433 | Each | 1 x 500 mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
0432 | Each | 1 x 100 mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
10018510 | Each | 3 x 18 mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|