Polyethyleneimine on silica beads, anion exchange resin, 20-40 mesh
Polyethyleneimine on silica beads, anion exchange resin, 20-40 mesh
Polyethyleneimine on silica beads, anion exchange resin, 20-40 mesh
Thermo Scientific Chemicals

Polyethyleneimine on silica beads, anion exchange resin, 20-40 mesh

For the separation of nucleotide mixtures | CAS: 9002-98-6 | ((C2H5N)zC2H4N)y(C2H5N)x | 43.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
250 mL
Catalog number 043302.AB
also known as 043302-AB
Price (USD)
47.70
Each
Quantity:
10 mL
Request bulk or custom format
Price (USD)
47.70
Each
Chemical Identifiers
CAS9002-98-6
Molecular Formula((C2H5N)zC2H4N)y(C2H5N)x
SMILES*-CCNCCN(-*)CCN-*
MDL NumberMFCD00084427
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown to orange to brown
Assay from Supplier's CofAMesh size 20-40
FormBeads
Polyethyleneimine is used for protein and nucleic acid precipitation, transfection assays and to pretreat filters to increase the binding of proteins. It can also complex heavy metal ions. An effective permeabilizer of gram negative bacteria. Crosslinked polyethyleneimine is used as an enzyme carrier. It is used as a membrane lubricant, useful in filtering frozen cell membranes and for the separation of nucleotide mixtures. Polyethyleneimine cellulose in small columns will separate cyclic nucleotides from related compounds at the same electrolyte concentration that is used for thin layer chromatography on PEI cellulose-coated plates. It is also used in separation medium for thin layer chromatography, paper chromatography and Low pressure liquid chromatography. Polyethylenimine on silica gel is used for thin layer chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only