Lithium tri-tert-butoxyaluminum hydride, nominally 1M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Lithium tri-tert-butoxyaluminum hydride, nominally 1M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium tri-tert-butoxyaluminum hydride, nominally 1M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
10 mL
Catalog number 043324.AD
also known as 043324-AD
Price (USD)/ Each
126.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
149.00 
Save 22.35 (15%)
-
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (USD)/ Each
126.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
149.00 
Save 22.35 (15%)
Add to cart
Lithium tri-tert-butoxyaluminum hydride, nominally 1M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Catalog number043324.AD
Price (USD)/ Each
126.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
149.00 
Save 22.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namealuminium(3+) tris(2-methylpropan-2-olate) lithium
Molecular FormulaC12H27AlLiO3
InChI KeyHTBVGZAVHBZXMS-UHFFFAOYSA-N
SMILES[Li].[Al+3].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-]
Molecular Weight (g/mol)253.27
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Gas Volumetric assayHydrogen evolution: 0.95-1.10M
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only