Thermo Scientific Chemicals

Acrylic acid, low water content, 99.5%, stab. with ca 200ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: 043359.AP
500 mL, Each
Thermo Scientific Chemicals

Acrylic acid, low water content, 99.5%, stab. with ca 200ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: 043359.AP
500 mL, Each
Quantity
Catalog number: 043359.AP
also known as 043359-AP
Price (USD)
Price: 47.60
Online price: 40.65
Your price:
Quantity
-

Chemical Identifiers

CAS
79-10-7
IUPAC Name
prop-2-enoic acid
Molecular Formula
C3H4O2
InChI Key
NIXOWILDQLNWCW-UHFFFAOYSA-N
SMILES
OC(=O)C=C
Water Content (Karl Fischer Titration)
≤0.1%
Appearance (Color)
Clear colorless
Form
Liquid
Assay from Supplier's CofA
≥99.4% (Assay: GC)
Comment
Diacrylic acid : ≤ 2000 ppm (Assay: LC)

Description

Acrylic acid acts as a precursor in the production of 3-hydroxypropionic acid. It is used in the preparation of water-absorbent resins. It reacts with alcohols to prepare the corresponding esters. Its esters are used as raw materials for synthetic resins, rubbers, coating adhesives, water-based paints, floor polishes and adhesives. It is also used to prepare forming homopolymers or copolymers by reacting with other monomers like acrylamides, acrylonitrile, vinyl, styrene, and butadiene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text