Thermo Scientific Chemicals

5-Hexyn-1-ol, 97%, Thermo Scientific Chemicals

Catalog number: 043370.06
5 g, Each
Thermo Scientific Chemicals

5-Hexyn-1-ol, 97%, Thermo Scientific Chemicals

Catalog number: 043370.06
5 g, Each
Quantity
Catalog number: 043370.06
also known as 043370-06
Price (USD)
Price: 92.20
Online price: 78.65
Your price:
Quantity
-

Chemical Identifiers

CAS
928-90-5
IUPAC Name
hex-5-yn-1-ol
Molecular Formula
C6H10O
InChI Key
GOQJMMHTSOQIEI-UHFFFAOYSA-N
SMILES
OCCCCC#C
Assay (GC)
> 96.0%

Description

5-Hexyn-1-ol is used as starting material for the preparation of terminal alkynes and lactones containing terminal alykyne groups. It is also used in the synthesis of cinnoline-fused cyclic enediyne, llycopodium alkaloids, (+)-nankakurine A and (+)-nankakurine B and 7-benzoyloxy-3-(2-nitrophenylseleno)-1,5-cyclodecadiyne.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text