1-Propanethiol, 98%
1-Propanethiol, 98%
1-Propanethiol, 98%
Thermo Scientific Chemicals

1-Propanethiol, 98%

CAS: 107-03-9 | C3H8S | 76.157 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 043420.AE
also known as 043420-AE
Price (EUR)
28,30
Each
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
28,30
Each
Add to cart
Chemical Identifiers
CAS107-03-9
IUPAC Namepropane-1-thiol
Molecular FormulaC3H8S
InChI KeySUVIGLJNEAMWEG-UHFFFAOYSA-N
SMILESCCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Colorless
FormLiquid
Retest Date1.4350-1.4390 @ 20?C
1-Propanethiol, is used widely in research areas. This compound has a wide range of applications including cell biology, materials science, nanoscience, self-assembled monolayers (SAMs), nanoelectronics, and contact printing. It is also used as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only