2-Propanethiol, 98%
2-Propanethiol, 98%
2-Propanethiol, 98%
2-Propanethiol, 98%
Thermo Scientific Chemicals

2-Propanethiol, 98%

CAS: 75-33-2 | C3H8S | 76.157 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 mL
100 mL
500 mL
產品號碼 043422.AC
亦稱為 043422-AC
價格 (TWD)
-
申請報價
Quantity:
25 mL
Request bulk or custom format
化學識別
CAS75-33-2
IUPAC Namepropane-2-thiol
Molecular FormulaC3H8S
InChI KeyKJRCEJOSASVSRA-UHFFFAOYSA-N
SMILESCC(C)S
檢視更多
規格Specification Sheet規格表
FormLiquid
Refractive Index1.4240-1.4280 @ 20?C
Appearance (Color)Colorless
Assay (GC)≥97.5%
2-Propanethiol Reacts with 3-chloro-propene to get 3-isopropylsulfanyl-propene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only