Aluminum foil, 0.5mm (0.02in) thick, annealed, Puratronic™, 99.9995% (metals basis)
Aluminum foil, 0.5mm (0.02in) thick, annealed, Puratronic™, 99.9995% (metals basis)
Aluminum foil, 0.5mm (0.02in) thick, annealed, Puratronic™, 99.9995% (metals basis)
Aluminum foil, 0.5mm (0.02in) thick, annealed, Puratronic™, 99.9995% (metals basis)
Thermo Scientific Chemicals

Aluminum foil, 0.5mm (0.02in) thick, annealed, Puratronic™, 99.9995% (metals basis)

CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 500 mm
50 x 50 mm
100 x 100 mm
Catalog number 043425.HP
also known as 043425-HP
Price (USD)/ Each
878.65
Special offer
Online exclusive
1,034.00
Save 155.35 (15%)
Add to cart
Quantity:
100 x 500 mm
Request bulk or custom format
Price (USD)/ Each
878.65
Special offer
Online exclusive
1,034.00
Save 155.35 (15%)
Add to cart
Chemical Identifiers
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
Molecular Weight (g/mol)26.98
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.9995% (metals basis)
Appearance (Color)Silvery white
FormFoil, 0.5mm (0.02in) thick, annealed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only