Polyethylene glycol 8,000
Polyethylene glycol 8,000
Polyethylene glycol 8,000
Polyethylene glycol 8,000
Thermo Scientific Chemicals

Polyethylene glycol 8,000

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
10 kg
Catalog number 043443.A3
also known as 043443-A3
Price (EUR)
137,00
Each
Quantity:
2 kg
Request bulk or custom format
Price (EUR)
137,00
Each
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Residue on ignition0.1 % max
Molecular Weight7000-9000 Average MW
pH4.5- 7.5 ( 5% solution at 25°C)
Trace Metals5 ppm, max (elemental impurities)
View more
Polyethylene glycol is used in the modification of therapeutic proteins and peptides to enhance its solubility. It is also used in the fabrication of bioactive and immuno isolating barriers for encapsulation of cells. In gas chromatography, it is used as a polar stationary phase. It is used as mold release agent and lubricant in fabricating elastomers. As a dispersant, it is used in toothpastes. It finds application in blood banking as a potentiator, which is used to detect antigens and antibodies. In pharmaceutical industry, it is used as tablet and capsule lubricant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only