Product Image
Thermo Scientific™
Dionex™ PC10 Postcolumn Delivery System Knitted Reaction Coils
Thermo Scientific™ Dionex™ PC10 Postcolumn Delivery System Knitted Reaction Coils is for use with the PC10 Postcolumn Delivery System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
043700375 μL
053640TS
also known as 053640
125 μL
Catalog number 043700
Price (MXN)
-
Capacity:
375 μL
Thermo Scientific™ Dionex™ PC10 Postcolumn Delivery System Knitted Reaction Coils is for use with the PC10 Postcolumn Delivery System.
The PC10 Postcolumn Pneumatic Delivery System ensures a constant, pulseless flow of reagent. It assures optimal flow by applying constant pressure, eliminating the need for a pump and its associated moving parts, labor and expense.
Specifications
Capacity375 μL
For Use With (Equipment)PC10 Post Column Delivery System, 4 mm
IncludesKnitted Reaction Coil For 4 mm System
OperationEnsures a constant, pulseless flow of reagent.
TypeReaction Coil
Unit SizeEach